Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062709.1 | XP_017918198.1 | 1 | 1024 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |