Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055782.1 | XP_017911271.1 | 3 | 1290 | 88 | |
GenBank mRNA | JO426454.1 | - | 1922 | 88 | ||
JO426457.1 | - | 1922 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |