Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172956.1 | NP_001166427.1 | 3 - 4 | 349 | 99 | |
XM_005007999.2 | XP_005008056.1 | 2 - 3 | 252 | 99 | ||
GenBank mRNA | AF033346.1 | - | 3 - 4 | 349 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Cavia porcellus Probe spans exons |