Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172996.1 | NP_001166467.1 | 1 - 2 | 145 | 72 | |
XM_005001384.3 | XP_005001441.1 | 2 - 3 | 294 | 72 | ||
GenBank mRNA | AF132929.1 | - | 1 - 2 | 145 | 72 | |
EB364243.1 | - | 1 - 2 | 94 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Cavia porcellus Probe spans exons |