Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172929.1 | NP_001166400.1 | 3 - 4 | 313 | 64 | |
GenBank mRNA | AF257213.1 | - | 3 - 4 | 313 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Cavia porcellus Probe spans exons |