Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172925.1 | NP_001166396.1 | 11 - 12 | 1480 | 81 | |
GenBank mRNA | AJ005111.1 | - | 11 - 12 | 1480 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Cavia porcellus Probe spans exons |