Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172936.1 | NP_001166407.1 | 5 - 6 | 567 | 88 | |
XM_005007749.3 | XP_005007806.1 | 5 - 6 | 612 | 88 | ||
XM_013142659.2 | XP_012998113.1 | 6 - 7 | 622 | 88 | ||
GenBank mRNA | M26936.1 | - | 5 - 6 | 567 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Cavia porcellus Probe spans exons |