Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina3k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173013.1 | NP_001166484.1 | 2 - 3 | 898 | 78 | |
XM_013144836.2 | XP_013000290.1 | 3 - 4 | 981 | 78 | ||
GenBank mRNA | M57269.1 | - | 2 - 3 | 898 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Cavia porcellus Probe spans exons |