Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173022.1 | NP_001166493.1 | 5 - 6 | 612 | 95 | |
GenBank mRNA | U09552.1 | - | 5 - 6 | 612 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Cavia porcellus Probe spans exons |