Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nptx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172728.1 | NP_001166199.1 | 3 - 4 | 1007 | 68 | |
GenBank mRNA | U13234.1 | - | 3 - 4 | 1007 | 68 | |
U13236.1 | - | 3 - 4 | 1015 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Cavia porcellus Probe spans exons |