Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172873.1 | NP_001166344.1 | 3 - 4 | 355 | 105 | |
GenBank mRNA | AY017306.1 | - | 3 - 4 | 355 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Cavia porcellus Probe spans exons |