Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173092.1 | NP_001166563.1 | 2 - 3 | 300 | 82 | |
GenBank mRNA | EB372154.1 | - | 2 - 3 | 300 | 82 | |
EU264005.1 | - | 2 - 3 | 300 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |