Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csn1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172894.1 | NP_001166365.1 | 4 - 5 | 147 | 85 | |
GenBank mRNA | X00374.1 | - | 4 - 5 | 147 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Cavia porcellus Probe spans exons |