Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csn1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172894.1 | NP_001166365.1 | 8 - 9 | 266 | 101 | |
GenBank mRNA | X00374.1 | - | 8 - 9 | 266 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Cavia porcellus Probe spans exons |