Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cma1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172932.1 | NP_001166403.1 | 4 - 5 | 633 | 93 | |
GenBank mRNA | AM851020.1 | - | 4 - 5 | 633 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |