Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172935.1 | NP_001166406.1 | 3 - 4 | 443 | 76 | |
XM_005000991.2 | XP_005001048.1 | 2 - 3 | 847 | 76 | ||
GenBank mRNA | DQ903937.1 | - | 2 - 3 | 374 | 76 | |
DQ903938.1 | - | 3 - 4 | 443 | 76 | ||
EB371635.1 | - | 3 - 4 | 211 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Cavia porcellus Probe spans exons |