Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173039.1 | NP_001166510.1 | 3 - 4 | 357 | 74 | |
XM_013147647.1 | XP_013003101.1 | 2 - 3 | 631 | 74 | ||
GenBank mRNA | EB370137.1 | - | 3 - 4 | 357 | 74 | |
EU827600.1 | - | 3 - 4 | 357 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Cavia porcellus Probe spans exons |