Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173073.1 | NP_001166544.1 | 10 - 11 | 1247 | 66 | |
XM_005004205.3 | XP_005004262.1 | 11 - 12 | 1376 | 66 | ||
XM_005004206.3 | XP_005004263.1 | 11 - 12 | 1336 | 66 | ||
XM_005004207.3 | XP_005004264.1 | 11 - 12 | 1376 | 66 | ||
GenBank mRNA | EU306867.1 | - | 10 - 11 | 1247 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |