Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173075.1 | NP_001166546.1 | 2 - 3 | 265 | 74 | |
GenBank mRNA | EU327786.1 | - | 2 - 3 | 265 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Cavia porcellus Probe spans exons |