Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172877.1 | NP_001166348.1 | 2 - 3 | 461 | 79 | |
GenBank mRNA | AY303774.1 | - | 2 - 3 | 461 | 79 | |
AY303775.1 | - | 2 - 3 | 415 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Cavia porcellus Probe spans exons |