Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173071.1 | NP_001166542.1 | 5 - 6 | 507 | 98 | |
GenBank mRNA | EB370475.1 | - | 5 - 6 | 497 | 98 | |
EF457998.1 | - | 5 - 6 | 507 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Cavia porcellus Probe spans exons |