Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173399.2 | NP_001166870.1 | 3 - 4 | 338 | 85 | |
GenBank mRNA | L04986.1 | - | 3 - 4 | 338 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |