Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173415.1 | NP_001166886.1 | 1 | 949 | 84 | |
XM_013156259.2 | XP_013011713.1 | 2 | 1453 | 84 | ||
XM_013156260.2 | XP_013011714.1 | 2 | 1204 | 84 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Cavia porcellus Both primers and probe map within a single exon |