Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172838.2 | NP_001166309.1 | 1 - 2 | 147 | 68 | |
GenBank mRNA | AB014340.1 | - | 1 - 2 | 148 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Cavia porcellus Probe spans exons |