Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pih1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135746.5 | NP_609590.1 | 2 - 3 | 307 | 100 | |
GenBank mRNA | AY061192.1 | - | 2 - 3 | 302 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |