Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DnaJ-H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273502.1 | NP_001260431.1 | 2 - 3 | 637 | 63 | |
NM_135761.3 | NP_609605.1 | 2 - 3 | 637 | 63 | ||
NM_165025.2 | NP_723785.1 | 3 - 4 | 1139 | 63 | ||
GenBank mRNA | BT150251.1 | - | 3 - 4 | 1139 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |