Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pect | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135769.3 | NP_609613.2 | 5 - 6 | 1041 | 63 | |
NM_165028.2 | NP_723789.2 | 5 - 6 | 1107 | 63 | ||
NM_165029.2 | NP_723790.2 | 5 - 6 | 1071 | 63 | ||
NM_165030.2 | NP_723791.2 | 5 - 6 | 1050 | 63 | ||
GenBank mRNA | BT003190.1 | - | 5 - 6 | 1065 | 63 | |
BT125823.1 | - | 5 - 6 | 818 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |