Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp28a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135850.3 | NP_609694.1 | 2 - 3 | 494 | 62 | |
GenBank mRNA | AY051533.1 | - | 2 - 3 | 470 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |