Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpr35B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135869.2 | NP_609713.1 | 2 | 392 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |