Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dao | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273533.1 | NP_001260462.1 | 4 - 5 | 2254 | 94 | |
NM_001273534.1 | NP_001260463.1 | 4 - 5 | 2254 | 94 | ||
NM_135898.4 | NP_609742.1 | 4 - 5 | 2254 | 94 | ||
GenBank mRNA | BT133421.1 | - | 4 - 5 | 2208 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |