Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpgamma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144358.3 | NP_001137830.2 | 10 - 11 | 2247 | 60 | |
NM_135958.5 | NP_609802.1 | 11 - 12 | 2290 | 60 | ||
NM_165169.5 | NP_723983.1 | 10 - 11 | 2247 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |