Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpgamma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144358.3 | NP_001137830.2 | 14 - 15 | 3220 | 68 | |
NM_135958.5 | NP_609802.1 | 15 - 16 | 3266 | 68 | ||
NM_165169.5 | NP_723983.1 | 14 - 15 | 3223 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |