Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpgamma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144358.3 | NP_001137830.2 | 15 - 16 | 3494 | 62 | |
NM_135958.5 | NP_609802.1 | 16 - 17 | 3540 | 62 | ||
NM_165169.5 | NP_723983.1 | 15 - 16 | 3497 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |