Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135969.4 | NP_609813.1 | 3 - 4 | 629 | 74 | |
NM_165194.3 | NP_724016.1 | 3 - 4 | 556 | 74 | ||
NM_165195.3 | NP_724017.1 | 3 - 4 | 876 | 74 | ||
NM_206002.2 | NP_995724.2 | 3 - 4 | 848 | 74 | ||
GenBank mRNA | AY051835.1 | - | 3 - 4 | 595 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |