Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-IIIc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135986.3 | NP_609830.2 | 3 - 4 | 704 | 66 | |
NM_165209.2 | NP_724042.1 | 3 - 4 | 704 | 66 | ||
GenBank mRNA | AY075458.1 | - | 3 - 4 | 703 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |