Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259129.2 | NP_001246058.1 | 2 | 1133 | 121 | |
NM_135998.3 | NP_609842.1 | 1 | 954 | 121 | ||
GenBank mRNA | AY094801.1 | - | 1 | 922 | 121 | |
BT124738.1 | - | 2 | 1133 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |