Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275240.1 | NP_001262169.1 | 1 - 2 | 206 | 66 | |
NM_141071.2 | NP_649328.1 | 1 - 2 | 206 | 66 | ||
GenBank mRNA | AY118790.1 | - | 1 - 2 | 206 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |