Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr47b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165826.3 | NP_725040.2 | 2 - 3 | 1104 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |