Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273961.1 | NP_001260890.1 | 3 - 4 | 894 | 66 | |
NM_001273962.1 | NP_001260891.1 | 3 - 4 | 668 | 66 | ||
NM_001273963.1 | NP_001260892.1 | 3 - 4 | 560 | 66 | ||
NM_001273964.1 | NP_001260893.1 | 3 - 4 | 1125 | 66 | ||
NM_001299387.1 | NP_001286316.1 | 3 - 4 | 1125 | 66 | ||
NM_078973.3 | NP_523697.1 | 4 - 5 | 824 | 66 | ||
NM_165833.3 | NP_725051.2 | 3 - 4 | 1125 | 66 | ||
GenBank mRNA | BT023883.1 | - | 3 - 4 | 1122 | 66 | |
BT126290.1 | - | 3 - 4 | 875 | 66 | ||
JN791442.1 | - | 3 - 4 | 548 | 66 | ||
JN791443.1 | - | 3 - 4 | 548 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |