Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tou | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103800.3 | NP_001097270.1 | 13 - 14 | 8564 | 71 | |
NM_001273969.1 | NP_001260898.1 | 13 - 14 | 8046 | 71 | ||
NM_001273970.1 | NP_001260899.1 | 12 - 13 | 8419 | 71 | ||
NM_078977.5 | NP_523701.3 | 13 - 14 | 8168 | 71 | ||
NM_165844.2 | NP_725062.2 | 2 - 3 | 540 | 71 | ||
GenBank mRNA | AY060335.1 | - | 2 - 3 | 540 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |