Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032236.3 | NP_001027407.2 | 4 - 5 | 1486 | 85 | |
NM_001032237.3 | NP_001027408.1 | 4 - 5 | 1805 | 85 | ||
NM_001103817.2 | NP_001097287.1 | 4 - 5 | 1805 | 85 | ||
NM_001259351.2 | NP_001246280.1 | 4 - 5 | 1805 | 85 | ||
NM_078988.4 | NP_523712.3 | 4 - 5 | 1324 | 85 | ||
NM_165904.3 | NP_725171.1 | 4 - 5 | 1805 | 85 | ||
NM_165905.4 | NP_725172.2 | 4 - 5 | 1324 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster Inventoried |