Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TppII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299447.1 | NP_001286376.1 | 4 - 5 | 1251 | 65 | |
NM_057899.3 | NP_477247.1 | 3 - 4 | 1163 | 65 | ||
NM_165966.2 | NP_725252.1 | 4 - 5 | 1517 | 65 | ||
GenBank mRNA | BT003260.1 | - | 3 - 4 | 1155 | 65 | |
BT044288.1 | - | 4 - 5 | 1468 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |