Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dh31-R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274022.2 | NP_001260951.1 | 10 - 11 | 1972 | 97 | |
NM_165979.3 | NP_725278.1 | 10 - 11 | 1972 | 97 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Drosophila melanogaster |