Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cad74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259885.1 | NP_001246814.1 | 8 - 9 | 5950 | 59 | |
NM_140716.2 | NP_648973.1 | 8 - 9 | 5664 | 59 | ||
NM_168724.2 | NP_730261.1 | 8 - 9 | 5679 | 59 | ||
GenBank mRNA | BT150474.1 | - | 8 - 9 | 5678 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |