Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eip74EF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259887.2 | NP_001246816.1 | 5 - 6 | 2790 | 65 | |
NM_168739.3 | NP_730286.3 | 5 - 6 | 2790 | 65 | ||
NM_168740.3 | NP_730287.1 | 5 - 6 | 2790 | 65 | ||
GenBank mRNA | M37082.1 | - | 5 - 6 | 3257 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |