Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vari | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273715.1 | NP_001260644.1 | 7 - 8 | 1610 | 81 | |
NM_001273716.1 | NP_001260645.1 | 6 - 7 | 1209 | 81 | ||
NM_165342.4 | NP_724288.3 | 7 - 8 | 1610 | 81 | ||
NM_165343.3 | NP_724289.2 | 6 - 7 | 1150 | 81 | ||
NM_206011.3 | NP_995733.2 | 7 - 8 | 1547 | 81 | ||
GenBank mRNA | BT056231.1 | - | 7 - 8 | 1546 | 81 | |
BT132965.1 | - | 7 - 8 | 1608 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |