Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CheB42b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206041.2 | NP_995763.1 | 1 - 2 | 250 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |