Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lig | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014503.2 | NP_001014503.1 | 3 - 4 | 2047 | 60 | |
NM_001169610.2 | NP_001163081.1 | 4 - 5 | 2040 | 60 | ||
NM_001169611.1 | NP_001163082.1 | 4 - 5 | 2064 | 60 | ||
NM_001169612.2 | NP_001163083.1 | 5 - 6 | 2106 | 60 | ||
NM_001259270.2 | NP_001246199.1 | 4 - 5 | 2040 | 60 | ||
NM_001259271.2 | NP_001246200.1 | 4 - 5 | 2040 | 60 | ||
NM_001273865.1 | NP_001260794.1 | 3 - 4 | 2047 | 60 | ||
NM_001273866.2 | NP_001260795.1 | 4 - 5 | 2040 | 60 | ||
NM_001316409.1 | NP_001303338.1 | 4 - 5 | 2040 | 60 | ||
NM_136504.5 | NP_610348.3 | 4 - 5 | 2040 | 60 | ||
NM_165586.5 | NP_724643.2 | 4 - 5 | 2109 | 60 | ||
NM_206056.5 | NP_995778.2 | 4 - 5 | 2395 | 60 | ||
GenBank mRNA | AF276964.1 | - | 3 - 4 | 2047 | 60 | |
AY069682.1 | - | 4 - 5 | 2064 | 60 | ||
BT100315.1 | - | 3 - 4 | 2013 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |