Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144147.2 | NP_001137619.1 | 10 - 11 | 4017 | 75 | |
NM_001273877.1 | NP_001260806.1 | 10 - 11 | 4017 | 75 | ||
NM_136542.3 | NP_610386.2 | 9 - 10 | 3057 | 75 | ||
NM_165609.3 | NP_724683.2 | 8 - 9 | 2111 | 75 | ||
NM_206058.2 | NP_995780.1 | 4 - 5 | 583 | 75 | ||
GenBank mRNA | BT033068.1 | - | 4 - 5 | 584 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Drosophila melanogaster |