Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136616.5 | NP_610460.1 | 1 - 2 | 980 | 54 | |
NM_206061.2 | NP_995783.1 | 1 - 2 | 417 | 54 | ||
GenBank mRNA | AY060801.1 | - | 1 - 2 | 387 | 54 | |
BT050564.1 | - | 1 - 2 | 948 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |